EMAIL THIS PAGE TO A FRIEND
Archives of toxicology
Investigation of the sensitising and cross-sensitising potential of textile dyes and beta-lactam antibiotics using a biphasic mice local lymph node assay.
PMID 19212757
Abstract
We used a modified protocol of the murine local lymph node assay (LLNA) to study the cross-sensitising potential of (a) textile dye disperse yellow 3 and its metabolite 2-amino-p-cresol, (b) two antibiotics, penicillin G and cefotiam. The test substances were applied in a biphasic manner, i.e. first on the shaved skin of the back followed by application on the dorsal side of the ears after 2 weeks. The end-points analysed included thickness and weight of an ear-biopsy, weight and cell number of the draining lymph node, and lymphocyte cell surface markers analysed by flow-cytometry. Disperse yellow 3 and its metabolite significantly altered the various end-points at both the tested concentrations (0.5 and 1%), thus demonstrating the sensitising potential of the two substances. The cross-sensitisation study showed significant modulation in the tested variables in the treated group as compared to the control, signifying cross-sensitisation potential of the two substances. Penicillin G and cefotiam showed significant changes in various end-points, pointing towards their sensitising potential. However, even at 50% concentration of the beta-lactams no significant change in any end-point indicating absence of cross-reactivity of the antibiotics was noticed. We conclude that a biphasic, modified protocol of the LLNA is a suitable approach to test for a cross-reactivity potential of two related compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
215686
Disperse Yellow 3, Dye content 30 %
C15H15N3O2
11344
Disperse Yellow 3, analytical standard
C15H15N3O2