EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
Synthesis and in-vitro antitumor activities of some mannich bases of 9-alkyl-1,2,3,4-tetrahydrocarbazole-1-ones.
PMID 19212985
Abstract
A novel series of 2-substituted aminomethyl-9-alkyl-1,2,3,4-tetrahydrocarbazole-1-ones 5a-q was synthesized via aminomethylation of 9-alkyl-1,2,3,4-tetrahydrocarbazole-1-ones 4a-e with hydrochlorides of the respective amines 6a-m. The structures of these newly synthesized compounds were characterized by (1)H-NMR, MS, and elemental analysis. All the compounds were tested for their cytotoxic activity in vitro against four human tumor cell lines including human non-small lung cancer cells (A549), human gastric adenocarcinoma (SGC), human colon cancer cell (HCT116), human myeoloid leukemia cells (K562), and one multi-drug resistant subline (KB-VCR). Most compounds showed moderate to potent cytotoxic activity against the tested cell lines. Preliminary mechanism research indicated that the most promising compound, 2-diethylaminomethyl-9-methyl-1,2,3,4-tetrahydrocarbazole-1-one 5c, exhibited a potential inhibitory effect against microtubule.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T12408
1,2,3,4-Tetrahydrocarbazole, 99%
C12H13N