EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
Facile, large-scale synthesis of dodecanethiol-stabilized Au38 clusters.
PMID 19215111
Abstract
It has long been a major challenge to achieve synthetic control over size and monodispersity of gold thiolate nanoclusters. Among the reported Aun thiolate clusters, Au38 has been shown to be particularly stable but was only obtained as a minor product in previous syntheses. In this work, we report a bulk solution synthetic method that permits large-scale, facile synthesis of truly monodisperse Au38 nanoclusters. This new method explores a two-phase ligand exchange process utilizing glutathione-capped Aun clusters as the starting material. The ligand exchange process with neat dodecanethiols causes gold core etching and secondary growth of clusters, and eventually leads to monodisperse Au38 clusters in high purity, which eliminates nontrivial postsynthetic separation steps. This method can be readily scaled up to synthesize Au38(SC12H25)24 in large quantities and thus makes the approach and Au38 nanoclusters of broad utility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44130
1-Dodecanethiol, purum, ≥97.0% (GC)
C12H26S
471364
1-Dodecanethiol, ≥98%
C12H26S