EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Stereoselective one-pot, three-component synthesis of 4-aryltetrahydropyran via Prins-Friedel-Crafts reaction.
PMID 19216514
Abstract
A diastereoselective one-pot, three-component Prins-Friedel-Crafts reaction was developed for the synthesis of 4-aryltetrahydropyran derivatives from the reaction of carbonyl compounds with homoallylic alcohol in the presence of arene promoted by boron trifluoride etherate.