EMAIL THIS PAGE TO A FRIEND
Journal of affective disorders
Select comorbid personality disorders and the treatment of chronic depression with nefazodone, targeted psychotherapy, or their combination.
PMID 19217168
Abstract
Individuals with chronic depression respond poorly to both medication and psychotherapy. The reasons for the poorer response, however, remain unclear. One potential factor is the presence of comorbid Axis II personality disorders (PDs), which occur at high rates among these patients. This study examines the moderating influence of co-occurring PDs, primarily in cluster C, among 681 chronically depressed adult outpatients who were randomly assigned to 12 weeks of treatment with nefazodone, a specialized psychotherapy for chronic depression, or their combination. At baseline, 50.4% (n=343) of patients met criteria for one or more Axis II disorders. Following 12 weeks of treatment, patients with comorbid PDs had statistically lower depression scores (M=12.2, SD=+9.2) than patients without comorbid PDs (M=13.5, SD=+8.7). There was no differential impact of a comorbid PD on responsiveness to medication versus psychotherapy. The results did not change when the data were analyzed using an intent-to-treat sample or when individual personality disorders were examined separately. Patients with severe borderline, antisocial, and schizotypal PDs were excluded from study entry; therefore, these data primarily apply to patients with cluster C PDs and may not generalize to other Axis II conditions. Comorbid Axis II disorders did not negatively affect treatment outcome and did not differentially affect response to psychotherapy versus medication. Treatment formulations for chronically depressed patients with certain PDs may not need to differ from treatment formulations of chronically depressed patients without co-occurring PDs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5536
Nefazodone hydrochloride, ≥98% (HPLC), solid
C25H32ClN5O2 · HCl