EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Atropisomeric small molecule Bcl-2 ligands: determination of bioactive conformation.
PMID 19217289
Abstract
The separation of atropisomeric conformers of 1,2,3,4-tetrahydroisoquinoline amide Bcl-2 ligands allowed the identification of the bioactive conformer which was subsequently confirmed by X-ray crystallography.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T13005
1,2,3,4-Tetrahydroisoquinoline, 95%
C9H11N