EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and biological evaluation of abietic acid derivatives.
PMID 19217699
Abstract
A series of C18-oxygenated derivatives of abietic acid were synthesized and evaluated for their cytotoxic, antimycotic, and antiviral activities. In general, the introduction of an aldehyde group at C18 did improve the resultant bioactivity, while the presence of an acid or alcohol led to less active compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00010
Abietic acid, technical, ~75% (GC)
C20H30O2