EMAIL THIS PAGE TO A FRIEND
The Journal of clinical psychiatry
Aripiprazole-induced agitation after clozapine discontinuation: a case report.
PMID 19222986
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1818
Molindone hydrochloride, ≥98% (HPLC), solid
C16H24N2O2 · HCl