Description
Molecular Formula
Add to Cart
HLTUD001C MISSION® Lenti microRNA Inhibitor, ath-miR416, Negative Control 1, Sequence from Arabidopsis thaliana with no homology to human and mouse gene sequences
HLTUD002C MISSION® Lenti microRNA Inhibitor, cel-miR-243-3p, Negative Control 2, Sequence from Caenorhabditis elegans with no homology to human and mouse gene sequences
HLTUD0172 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-1302
HLTUD1194 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4317
HLTUD1196 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4321
HLTUD1197 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4323
HLTUD1198 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4324
HLTUD1199 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4256
HLTUD1200 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4257
HLTUD1201 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4258
HLTUD1202 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4259
HLTUD1204 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4253
HLTUD1205 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4251
HLTUD1206 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4254
HLTUD1207 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4255
HLTUD1208 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4252
HLTUD1209 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4325
HLTUD1210 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4326
HLTUD1212 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4261
HLTUD1213 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4265
HLTUD1214 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4266
HLTUD1215 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4267
HLTUD1216 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4262
HLTUD1217 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-2355-5p
HLTUD1219 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4269
HLTUD1220 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4263
HLTUD1221 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4264
HLTUD1222 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4270
HLTUD1223 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4271
HLTUD1224 MISSION® Lenti microRNA Inhibitor, Human, hsa-miR-4272