EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
Quantification of tryptophan transport and metabolism in lung tumors using PET.
PMID 19223408
Abstract
Abnormal tryptophan metabolism catalyzed by indoleamine 2,3-dioxygenase may play a prominent role in tumor immunoresistance in many tumor types, including lung tumors. The goal of this study was to evaluate the in vivo kinetics of alpha-(11)C-methyl-l-tryptophan (AMT), a PET tracer for tryptophan metabolism, in human lung tumors. Tracer transport and metabolic rates were evaluated in 18 lesions of 10 patients using dynamic PET/CT with AMT. The kinetic values were compared between tumors and unaffected lung tissue, tested against a simplified analytic approach requiring no arterial blood sampling, and correlated with standardized uptake values (SUVs) obtained from (18)F-FDG PET/CT scans. Most non-small cell lung cancers (NSCLCs) showed prolonged retention of AMT, but 3 other lesions (2 benign lesions and a rectal cancer metastasis) and unaffected lung tissue showed no such retention. Transport and metabolic rates of AMT were substantially higher in NSCLCs than in the other tumors and unaffected lung tissue. A simplified analytic approach provided an excellent estimate of transport rates but only suboptimal approximation of tryptophan metabolic rates. (18)F-FDG SUVs showed a positive correlation with AMT uptake, suggesting higher tryptophan transport and metabolism in tumors with higher proliferation rates. Prolonged retention of AMT in NSCLCs suggests high metabolic rates of tryptophan in these tumors. AMT PET/CT may be a clinically useful molecular imaging method for personalized cancer treatment by identifying and monitoring patients who have increased tumor tryptophan metabolism and are potentially sensitive to immunopharmacotherapy with indoleamine 2,3-dioxygenase inhibitors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8377
α-Methyl-DL-tryptophan, crystalline
C12H14N2O2