EMAIL THIS PAGE TO A FRIEND
Diabetologia
A common variant in PNPLA3, which encodes adiponutrin, is associated with liver fat content in humans.
PMID 19224197
Abstract
It has recently been suggested that the rs738409 G allele in PNPLA3, which encodes adiponutrin, is strongly associated with increased liver fat content in three different ethnic groups. The aims of the present study were as follows: (1) to try to replicate these findings in European individuals with quantitative measures of hepatic fat content; (2) to study whether the polymorphism influences hepatic and adipose tissue insulin sensitivity; and (3) to investigate whether PNPLA3 expression is altered in the human fatty liver. We genotyped 291 Finnish individuals in whom liver fat had been measured using proton magnetic resonance spectroscopy. Hepatic PNPLA3 expression was measured in 32 participants. Hepatic and adipose tissue insulin sensitivities were measured using a euglycaemic-hyperinsulinaemic (insulin infusion 0.3 mU kg(-1) min(-1)) clamp technique combined with infusion of [3-(3)H]glucose in 109 participants. The rs738409 G allele in PNPLA3 was associated with increased quantitative measures of liver fat content (p = 0.011) and serum aspartate aminotransferase concentrations (p = 0.002) independently of age, sex and BMI. Fasting serum insulin and hepatic and adipose tissue insulin sensitivity were related to liver fat content independently of genotype status. PNPLA3 mRNA expression in the liver was positively related to obesity (r = 0.62, p < 0.0001) and to liver fat content (r = 0.58, p = 0.025) in participants who were not morbidly obese (BMI < 40 kg/m(2)). A common variant in PNPLA3 increases the risk of hepatic steatosis in humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62287 Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
L4777
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
62288 Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
62301 Lipase from Aspergillus niger, powder (fine), ~200 U/g
L4277 Lipase from Aspergillus oryzae, ≥20,000 U/g
62285 Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
02569 Lipase from Candida antarctica, lyophilized, powder, beige, ~0.3 U/mg
L1754 Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
L8525 Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
62316 Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
62302 Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
L3170
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
L8906 Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
L9031 Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
62298 Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
L0382 Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
62309 Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
L9518
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
62310 Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
62305 Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
L3001 Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
62306 Lipase from wheat germ, ~0.1 U/mg
73940 Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg