EMAIL THIS PAGE TO A FRIEND
BJU international
A review of the use of histrelin acetate in the treatment of prostate cancer.
PMID 19228148
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2761
[des-Gly10, D-His(Bzl)6]-LH-RH ethylamide, ≥97% (HPLC), powder
C66H86N18O12