EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Sphingomyelin synthase 2 is palmitoylated at the COOH-terminal tail, which is involved in its localization in plasma membranes.
PMID 19233134
Abstract
Sphingomyelin synthase (SMS) is an enzyme that catalyzes the transfer of phosphocholine from phosphatidylcholine to ceramide for sphingomyelin synthesis. Here, we show that SMS2 is palmitoylated at cysteine residues via thioester bonds in the COOH-terminal cytoplasmic tail. [3H]palmitic acid labeling of SMS1 or SMS2-overexpressing HEK293 cells revealed that SMS2, but not SMS1, is palmitoylated. Site-directed mutagenesis of cysteine residues to alanine ones indicated that the COOH-terminal cysteine cluster of the enzyme is palmitoylated. Mutation of all potential palmitoylation sites resulted in a dramatic reduction in the plasma membrane distribution of SMS2, whereas it did not affect the in vitro enzyme activity. These results suggested that this posttranslational modification is important for determination of the subcellular localization of SMS2.