EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Facile synthesis of beta-tribromomethyl and dibromomethylenated nitroalkanes via conjugate addition of bromoform to nitroalkenes.
PMID 19239256
Abstract
Addition of bromoform to conjugated nitroalkenes in the presence of Mg provided beta-tribromomethyl nitroalkanes in good to excellent yields and diastereoselectivity. These novel Michael adducts, formed under radical conditions, underwent elimination of HBr in the same pot under reflux to afford beta-dibromomethylenated nitroalkanes in good yield. Alternatively, a one-pot high yielding synthesis of the dibromides was possible under anionic conditions via LDA mediated addition of bromoform to nitroalkenes.