EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Synthesis of thiol-linked neoglycopolymers and thermo-responsive glycomicelles as potential drug carrier.
PMID 19240872
Abstract
Homopolymer and block copolymer bearing carbohydrate side chain functionality were obtained by grafting glucothiose onto alkene functional scaffolds via a thiol-ene click reaction and the resulting copolymer was used to form thermo-responsive micelles as a potential drug carrier.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471801
4-Pentenoic anhydride, 98%
C10H14O3