EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
TOF-S-SIMS molecular depth profiling of organic bilayers using mechanical wear test methodology.
PMID 19241066
Abstract
Recent publications on static secondary ion mass spectrometry (S-SIMS) focus on molecular depth profiling by using polyatomic or ultra-low energy monoatomic projectiles. Since their applicability depends on the relationship between the ion yield and the depth, which is hard to obtain without extensive studies, a combination of a wear test method with S-SIMS surface analysis was performed in the current study. Using this non-sputtering procedure, the relation between the signal intensity and the local concentration remains in principle the same as that at the surface (which is easy to determine). Mechanical erosion was successfully applied to expose sub-surface material from organic multilayers. Through surface analysis with S-SIMS on the gradually exposed deeper planes, molecular depth profiles could be obtained. The study was conducted on a model system relevant to offset printing, consisting of two polymer layers, containing dyes and a surfactant, cast on an Al substrate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
228842
Ethyl Violet, cationic triarylmethane dye
C31H42ClN3