EMAIL THIS PAGE TO A FRIEND
Dental materials journal
Effects of visible light-irradiated camphorquinone and 9-fluorenone on murine oral mucosa.
PMID 19241689
Abstract
The purpose of this study was to evaluate the histopathological effects of camphorquinone (CQ) and 9-fluorenone (9F) with or without visible light (VL) irradiation on the oral mucous membranes of mice. VL irradiation resulted in a higher degree of tissue damage after CQ or 9F application, particularly the latter. Necrosis and apoptosis were responsible for the tissue damage after application of either agent in the presence of VL irradiation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F1506
9-Fluorenone, 98%
C13H8O