EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
A novel compound isolated from the peels of Citrus changshan-huyou.
PMID 19244751
Abstract
Five compounds, huyouyisu (1), vomifoliol (2), isoferulic acid (3), 1,2,3-trihydroxyphenol (4) and p-hydroxy-phenol (5), were isolated from the peels of Citrus changshan-huyou Y. B. Chang for the first time by utilizing repeated column chromatography on silica gel. Among them, huyouyisu (1) is a new compound. The structure was elucidated by using 1D and 2D spectra.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
103012
3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 97%
C10H10O4