EMAIL THIS PAGE TO A FRIEND
Molecular diversity
Organic synthesis in water: a green protocol for the synthesis of 2-(cyclohexylamino)-3- aryl- indeno[1,2-b]furan-4-ones.
PMID 19252994
Abstract
Aldehydes, 1,3-indandione and cyclohexylisocyanide undergo smooth coupling-cyclization in water to produce the corresponding 2-(cyclohexylamino)-3-aryl- indeno [1,2-b] furan-4-ones in good yields. Water was used as a solvent to avoid the use of other highly toxic and environmentally unfavorable solvents for this synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2002
1,3-Indandione, 97%
C9H6O2