EMAIL THIS PAGE TO A FRIEND
Luminescence : the journal of biological and chemical luminescence
Flow-injection chemiluminescence method for the determination of naphazoline hydrochloride and oxymetazoline hydrochloride.
PMID 19253271
Abstract
A sensitive and simple flow-injection chemiluminescence (FI-CL) method, which was based on the CL intensity generated from the redoxreaction of potassium permanganate (KMnO4)-formaldehyde in vitriol (H2SO4) medium, has been developed, validated and applied for the determination of naphazoline hydrochloride and oxymetazoline hydrochloride. Besides oxidants and sensitizers, the effect of the concentration of H(2)SO(4), KMnO4 and formaldehyde was investigated. Under the optimum conditions, the linear range was 1.0 x 10(-2)-7.0 mg/L for naphazoline hydrochloride and 5.0 x 10(-2)-10.0 mg/L for oxymetazoline hydrochloride. During seven repeated inter-day and intra-day precision tests of 0.1, 1.0 and 10.0 mg/L samples, the relative standard deviations all corresponded to reference values. The detection limit was 8.69 x 10(-3) mg/L for naphazoline hydrochloride and 3.47 x 10(-2) mg/L for oxymetazoline hydrochloride (signal-to-noise ratio < or = 3). This method has been successfully implemented for the determination of naphazoline hydrochloride and oxymetazoline hydrochloride in pharmaceuticals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5504
Naphazoline hydrochloride
C14H14N2 · HCl