EMAIL THIS PAGE TO A FRIEND
Revista de enfermeria (Barcelona, Spain)
[Biological changes in the skin].
PMID 1925336
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0891
Flumethasone pivalate
C27H36F2O6