EMAIL THIS PAGE TO A FRIEND
TSitologiia i genetika
[Mechanisms of action of plant resistance inductor beta-aminobutyric acid in Allium cepa].
PMID 19253749
Abstract
The effects of beta-aminobutiric acid as an inductor of plant resistance was studied. The influence of BABA pretreatment on A. cepa plants of three different pathogen-resistant varieties was investigated by fluorescence microscopic methods. It was determined that BABA is able to prime the plant callose accumulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
757454
(S)-3-Aminobutyric acid, 97%
C4H9NO2
A44207
3-Aminobutanoic acid, 97%
C4H9NO2