EMAIL THIS PAGE TO A FRIEND
Organic letters
Protecting group and solvent control of stereo- and chemoselectivity in glucal 3-carbamate amidoglycosylation.
PMID 19254041
Abstract
In the Rh(2)(OAc)(4)-catalyzed amidoglycosylation of glucal 3-carbamates, anomeric stereoselectivity and the extent of competing C3-H oxidation depend on the 4O and 6O protecting groups. Acyclic protection permits high alpha-anomer selectivity with further improvement in less polar solvents, while electron-withdrawing protecting groups limit C3-oxidized byproducts. Stereocontrol and bifurcation between alkene insertion and C3-H oxidation reflect an interplay of conformational, stereoelectronic, and inductive factors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4670
D-Mannosamine hydrochloride
C6H13NO5 · HCl