EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Characterization of macrolide efflux pump mef subclasses detected in clinical isolates of Streptococcus pyogenes isolated between 1999 and 2005.
PMID 19258262
Abstract
The macrolide efflux mechanism of resistance, mef, was characterized in community-acquired respiratory tract infections with Streptococcus pyogenes. Fifty-four (4.6%) M phenotype isolates were screen tested as negative for mef(A). Of these 54 isolates, 5 (0.4%), 27 (2.3%), and 1 (0.1%) were considered to be mef(I) positive, a novel mosaic variant of mef, or a novel subclass of mef, respectively. This study shows (i) the definitive presence of mef(E) in S. pyogenes and its global distribution, (ii) the presence of a mosaic variant of mef composed of mef(A) and mef(E), (iii) the previously undescribed presence of mef(I) in S. pyogenes, and (iv) the presence of a novel subclass of mef in S. pyogenes.