EMAIL THIS PAGE TO A FRIEND
The Pediatric infectious disease journal
Genetic susceptibility to respiratory syncytial virus bronchiolitis in preterm children is associated with airway remodeling genes and innate immune genes.
PMID 19258923
Abstract
Prematurity is a risk factor for severe respiratory syncytial virus bronchiolitis. We show that genetic factors in innate immune genes (IFNA13, IFNAR2, STAT2, IL27, NFKBIA, C3, IL1RN, TLR5), in innate and adaptive immunity (IFNG), and in airway remodeling genes (ADAM33 and TGFBR1), affect disease susceptibility to a different extent in preterm children, born with underdeveloped lungs, than in term children.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10478 CompoZr® Knockout ZFN Kit, Human IFNA13 (NM_006900)