EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Analysis of volatile oil in pre and post processed pieces of Eriobotrya japonica by GC-MS].
PMID 19260269
Abstract
To analyze the chemical components of the volatile oil in processed pieces of Eriobotrya japonica by GC-MS. The volatile oil was obtained by steam distillation. The amount of the components from the volatile oil were determinated by area normalization method. The separated components were identified by GC-MS. The components of volatile oil in pre and post processed pieces of Eriobotrya japonica were different. The methods is reliable, stable and has good repeatability. It will provide the certain scientific methods for further evaluation quality of Eriobotrya japonica.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442505
Camphene, analytical standard
C10H16
456055
Camphene, 95%
C10H16