EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Triterpenoids and steroids of root of Rubus biflorus].
PMID 19260277
Abstract
To investigate the chemical constituents of root of Rubus biflorus. Isolation and purification were carried out by silica gel column chromatography and recrystallization and so on. Structural determination of the pure compounds was based on physico-chemical properties and various spectral data analysis (1H-NMR, 13C-NMR, IR, EI-MS). Five compounds were isolated and identified as follows: lupeol acetate (I), corosolic acid (II), oleanic acid (III), 2alpha-hydroxyoleanolic acid (IV), sitosterol (V). All these compounds are isolated from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0549
Corosolic acid from Lagerstroemia speciosa, ≥98% (HPLC)
C30H48O4