EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Determination of eleutheroside B in antifatigue fraction of Acanthopanax senticosus by HPLC].
PMID 19260316
Abstract
To develop an HPLC method for the determination of eleutheroside B in As1 (the extract fraction from Acanthopanax senticosus, which has a good effect of antifatigue). The antifatigue effect of Asl was evaluated by mice burden swimming testing. One compound was isolated by silica gel, Sephadex LH-20 column chromatography and preparative high performance liquid chromatography. The structure was identified by physicochemical properties and spectral evidences. Eleutheroside B in Asl was determined by HPLC. Chromatographic conditions included Diamonsil C18 column (4.6 mm x 250 mm, 5 microm) and the mobile phase consisting of a mixture of acetonitrile-water-ethanoic acid (10: 90: 0.01). The UV detection wavelength was set at 344 nm. As1 showed an excellent antifatigue activity; One compound was isolated from As1 and its structure was identified as Eleutheroside B; The calibration cure was linear in the range of 0.104-20.8 microg (r = 0.9999), the average recovery was 97.68%, RSD 1.4% (n=6). This HPLC method is simple,accurate and reliable.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90974
Eleutheroside B, analytical standard
C17H24O9