EMAIL THIS PAGE TO A FRIEND
Organic letters
Highly enantioselective synthesis of nitrocyclopropanes via organocatalytic conjugate addition of bromomalonate to alpha,beta-unsaturated nitroalkenes.
PMID 19260703
Abstract
Highly enantioselective synthesis of nitrocyclopropanes was achieved via the organocatalytic conjugate addition of dimethyl bromomalonate to nitroalkenes and the consequent intramolecular cyclopropanation. 6'-Demethyl quinine was found to be the efficient catalyst. Excellent enantioselectivities, diastereoselectivities, and good yields were obtained for a variety of aryl or heteroaryl nitroethylenes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
359890
Dimethyl bromomalonate, technical grade, 90%
C5H7BrO4