EMAIL THIS PAGE TO A FRIEND
Reproduction (Cambridge, England)
Pre-fertilization zona pellucida hardening by different cross-linkers affects IVF in pigs and cattle and improves embryo production in pigs.
PMID 19261833
Abstract
Zona pellucida (ZP) hardening (resistance to proteolysis) has been classically identified as a post-fertilization event that contributes to the block to polyspermy. Di-(N-succinimidyl)-3,3'-dithiodipropionate (DSP), a permeable amine-reactive cross-linker, was recently shown to induce pre-fertilization ZP hardening and to improve porcine IVF productivity. The objectives of this study were to investigate i) how DSP affects pre-fertilization ZP hardening and IVF in cattle, ii) if a non-permeable amine-reactive cross-linker such as bis(sulfosuccinimidyl) suberate (BS3) affects ZP hardening and IVF in cattle and pigs, and iii) whether DSP or BS3, if improvement in IVF productivity was demonstrated in either species, affects in vitro embryo development. Bovine and porcine in vitro matured oocytes were incubated with the cross-linkers (0.06, 0.3, and 0.6 mg/ml) for 30 min. Then they were subjected to ZP digestion or IVF. In cattle, both DSP and BS3 induced ZP hardening and decreased the penetration rate, although monospermy, penetration, or male pronuclear formation was not affected. In pigs, BS3 treatment induced ZP hardening, decreased penetration and male pronuclear formation, and increased monospermy. IVF productivity only improved when porcine oocytes were exposed to DSP. When porcine zygotes derived from this treatment were further cultured in vitro, the cleavage and blastocyst formation rates increased. These results support the idea that mechanisms involved in the prevention of polyspermic fertilization in cattle and pigs have different efficiencies, and ZP hardening induced by DSP cross-linker may be useful for improving porcine embryo production.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3669
3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), powder
C14H16N2O8S2