EMAIL THIS PAGE TO A FRIEND
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine
High specific activity tritiation of TCP and BTCP.
PMID 19264496
Abstract
Methods are described to synthesize and characterize tritium labelled TCP and BTCP at high specific activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B138
BTCP hydrochloride
C19H25NS · HCl