EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
Alterations in nicotinic α4β2 receptor binding in vascular dementia using ¹²³I-5IA-85380 SPECT: comparison with regional cerebral blood flow.
PMID 19269714
Abstract
To investigate differences in distribution of α4β2 subtypes of nicotinic acetylcholine receptors (nAChRs) using the ligand ¹²³I-5-Iodo-3-[2(S)-2-azetidinylmethoxy] pyridine (5IA-85380) and single photon emission computed tomography (SPECT) in subjects with vascular dementia and age-matched controls. ¹²³I-5IA-85380 binding was compared to corresponding regional cerebral blood flow (rCBF) changes in the same subjects. Thirty subjects (14 vascular dementia and 16 controls) underwent ¹²³I-5IA-85380 and rCBF ((99m)Tc-exametazime) SPECT scanning. Image analysis was performed on voxel basis using statistical parametric mapping (SPM2). Compared to controls, reductions in relative ¹²³I-5IA-85380 uptake were identified in dorsal thalamus and right caudate in vascular dementia. Increase in scaled ¹²³I-5IA-85380 uptake in cuneus was also demonstrated in vascular dementia relative to controls. Perfusion deficits in anterior cingulate were apparent in the patient group and did not appear to be associated with ¹²³I-5IA-85380 changes. Reduced ¹²³I-5IA-85380 uptake in vascular dementia was confined to sub-cortical regions, unlike the cortical reductions previously described in Alzheimer's disease. Elevation of normalised ¹²³I-5IA-85380 uptake in cuneus in vascular dementia could be a compensatory response to reduced cholinergic activity in dorsal thalamus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000227 Meso-rich exametazime, European Pharmacopoeia (EP) Reference Standard
C13H28N4O2