EMAIL THIS PAGE TO A FRIEND
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Nature of histamine transport in neonatal rat cultured type 1 astrocytes--organic cation transporters are not involved.
PMID 19271125
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323764
1,1′-Diethyl-2,2′-cyanine iodide, 97%
C23H23IN2