EMAIL THIS PAGE TO A FRIEND
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Characterization of histamine-induced relaxation in pre-contracted rat aorta.
PMID 19271142
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1915
Chloropyramine hydrochloride, analytical standard, for drug analysis
C16H20ClN3 · HCl