EMAIL THIS PAGE TO A FRIEND
Journal of obstetrics and gynaecology : the journal of the Institute of Obstetrics and Gynaecology
Ligneous cervicitis: an unusual cause of post-coital bleeding in a postmenopausal woman.
PMID 19274564