EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Use of the sulfide mineral pyrite as electrochemical sensor in non-aqueous solutions: potentiometric titration of weak acids in acetonitrile, propionitrile and benzonitrile.
PMID 19276604
Abstract
Natural monocrystalline pyrite as a new indicator electrode for the potentiometric titration of weak acids in acetonitrile, propionitrile and benzonitrile was studied. The investigated electrode showed a linear dynamic response for p-toluenesulfonic acid concentrations in the range from 0.1 to 0.001 M, with a Nernstian slope of 74 mV per decade. Sodium methylate, potassium hydroxide and tetrabutylammonium hydroxide (TBAH) proved to be very suitable titrating agent for this titration. The response time was less than (11 s) and the lifetime of the electrode is long. The advantages of the electrode are log-term stability, fast response, and reproducibility, while the sensor is easy to prepare and of low cost.
Related Materials