EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Determination of total retronecine esters-type hepatotoxic pyrrolizidine alkaloids in plant materials by pre-column derivatization high-performance liquid chromatography.
PMID 19277972
Abstract
A pre-column derivatization high-performance liquid chromatography method with diode array detection was developed and validated to determine the total retronecine esters-type hepatotoxic pyrrolizidine alkaloids (RET-HPAs) in herbs. The RET-HPAs reacted with o-chloranil in methanolic solution heated for 3 h, and an oxidative derivative was produced that could be detected at a maximal absorption of 223 nm. The analysis was performed using a C18 column with an isocratic elution of methanol and aqueous 0.01% triethylamine (adjusted to pH 4 with formic acid), and the detection was carried out with DAD at 223 nm. The validation of the method included linearity, sensitivity, recovery and stability. It showed a good linear regression (r(2) > 0.9900) in the range of 2.5-250 microM with a limit of detection (S/N = 3) of 0.5 microM. The method provided desirable repeatability with overall intra- and inter-day variations of less than 4.6%. The obtained recoveries for both of the extraction and derivatization process were between 94.6 and 100.7% (n = 3).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6601
3,4,5,6-Tetrachloro-1,2-benzoquinone, 97%
C6Cl4O2
32328
Retronecine, analytical standard
C8H13NO2