EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Applicability of LC/MS/MS assay for 6-methoxy-2-napthylacetic acid to support the bioequivalence study of nabumetone-comments on the research work of Patel et al. (2008).
PMID 19277989