EMAIL THIS PAGE TO A FRIEND
The Journal of Neuroscience
Dopamine and amphetamine rapidly increase dopamine transporter trafficking to the surface: live-cell imaging using total internal reflection fluorescence microscopy.
PMID 19279270