EMAIL THIS PAGE TO A FRIEND
Acta biochimica et biophysica Sinica
The effect of bafilomycin A1 and protease inhibitors on the degradation and recycling of a Class 5-mutant LDLR.
PMID 19280064
Abstract
The low-density lipoprotein receptor (LDLR) mediates cholesterol homeostasis through endocytosis of lipoprotein particles, particularly low-density lipoproteins (LDLs). Normally, the lipoprotein particles are released in the endosomes and the receptors recycle to the cell surface. Familial hypercholesterolemia (FH) is an autosomal dominant disease caused by mutations in the gene encoding the LDLR. These mutations are divided into five functional classes where Class 5 mutations encode receptors that suffer from ligand-induced degradation and recycling deficiency. The aim of this study was to investigate whether it is possible to prevent the fast ligand-induced degradation of Class 5-mutant LDLR and to restore its ability to recycle to the cell surface. E387K is a naturally occurring Class 5 mutation found in FH patients, and in the present study, we used Chinese hamster ovary cells transfected with an E387K-mutant LDLR. Abrogation of endosomal acidification by adding bafilomycin A1 or addition of the irreversible serine protease inhibitors, 4-(2-aminoethyl)-benzenesulfonyl fluoride (AEBSF) and 3,4-dichloroisocoumarin (DCI), prevented the degradation of the E387K-mutant LDLR. However, the undegraded receptor did not recycle to the cell surface in the presence of LDL. Unexpectedly, AEBSF caused aggregation of early endosome antigen-1- positive endosomes and the intracellular trapped LDLR co-localized with these aggregated early endosomes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7910
3,4-Dichloroisocoumarin, serine protease inhibitor
C9H4Cl2O2