EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Ganoderic acid DM: anti-androgenic osteoclastogenesis inhibitor.
PMID 19289282
Abstract
Prostate cancer is the most common cancer in men in Western countries, with a high incidence of bone metastasis. Ganoderic acid DM, with 5alpha-reductase inhibitory and androgen receptor (AR) binding activity, isolated from the ethanol extracts of Ganoderma lucidum, can inhibit prostate cancer cell growth and block osteoclastogenesis.