EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Zocor Forte (simvastatin) has a neuroprotective effect against LPS striatal dopaminergic terminals injury, whereas against MPP+ does not.
PMID 19292984
Abstract
Due to their potential role in preventing further deterioration of Parkinson's disease, anti-inflammatory strategies have attracted great interest. In this context, some studies point out the possible protective effect of anti-inflammatory compounds against the in vivo degeneration of dopaminergic neurons produced by lipopolysaccharide (LPS)-induced inflammatory processes and others. We have investigated the effect of the treatment of Zocor Forte (simvastatin) in LPS and 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) neurodegenerative models to identify neuroprotective drugs for Parkinson's disease. We have perfused different concentrations of LPS or 1 mM 1-methyl-4-phenylpyridinium ion (MPP+) in the rat's striatum, 24 h after implanting a brain microdialysis probe, both with and without Zocor Forte (simvastatin) treatment. Results show that LPS perfusion produced a decrease in the basal release of dopamine. Forty-eight hours after implanting the probe, we have perfused 1 mM MPP+ to check the integrity of the dopaminergic terminals present around the cannula. Our model to study toxicity in the striatal dopaminergic terminals suggests that Zocor Forte (simvastatin) could prevent the neurotoxic damage produced by LPS, but not that produced by MPP+.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
416835
4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride, technical grade
C11H13N · HCl