EMAIL THIS PAGE TO A FRIEND
ACS chemical biology
Expanding the repertoire of fluorescent calcium sensors.
PMID 19298092
Abstract
Fluorescent indicators for calcium are incredibly powerful because they enable researchers to watch the movement of calcium ions in real time in living cells. The popular small molecule indicator Indo-1 has now been targeted to a defined location, namely, the nucleus of muscle cells, using SNAP-tag technology. This combination of a chemical probe with genetic targeting expands the available options for measuring local calcium events in cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57180
Indo 1, for fluorescence, ≥80.0% (HPLC)
C32H26K5N3O12