EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
A quantitative NMR protocol for the simultaneous analysis of atropine and obidoxime in parenteral injection devices.
PMID 19299099
Abstract
A rapid selective and accurate quantitative (1)H NMR method was developed for the simultaneous analysis of obidoxime chloride and atropine sulfate, the active components in parenteral injection devices (PID) used for the emergency treatment of poisoning by toxic organophosphates. The spectra were acquired in 90% H(2)O-10% D(2)O using sodium 3-(trimethylsilyl)-1-propane sulfonate hydrate as the internal standard. Both synthetic mixtures and dosage forms were assayed. The results were compared with those obtained from a reported HPLC method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
178837
3-(Trimethylsilyl)-1-propanesulfonic acid sodium salt, 97%
C6H15NaO3SSi