EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Effect of estrogenic and antiestrogenic triphenylethylene derivatives on progesterone and estrogen receptors levels of MCF-7 cells.
PMID 1930303
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T82805
Triphenylethylene, 99%
C20H16