EMAIL THIS PAGE TO A FRIEND
Phytomedicine : international journal of phytotherapy and phytopharmacology
Hypoglycemic activity of a novel anthocyanin-rich formulation from lowbush blueberry, Vaccinium angustifolium Aiton.
PMID 19303751
Abstract
Blueberry fruits are known as a rich source of anthocyanin components. In this study we demonstrate that anthocyanins from blueberry have the potency to alleviate symptoms of hyperglycemia in diabetic C57b1/6J mice. The anti-diabetic activity of different anthocyanin-related extracts was evaluated using the pharmaceutically acceptable self-microemulsifying drug delivery system: Labrasol. Treatment by gavage (500 mg/kg body wt) with a phenolic-rich extract and an anthocyanin-enriched fraction formulated with Labrasol lowered elevated blood glucose levels by 33 and 51%, respectively. The hypoglycemic activities of these formulae were comparable to that of the known anti-diabetic drug metformin (27% at 300 mg/kg). The extracts were not significantly hypoglycemic when administered without Labrasol, demonstrating its bio-enhancing effect, most likely due to increasing the bioavailability of the administered preparations. The phenolic-rich extract contained 287.0+/-9.7 mg/g anthocyanins, while the anthocyanin-enriched fraction contained 595+/-20.0 mg/g (cyanidin-3-glucoside equivalents), as measured by HPLC and pH differential analysis methods. The greater hypoglycemic activity of the anthocyanin-enriched fraction compared to the initial phenolic-rich extract suggested that the activity was due to the anthocyanin components. Treatment by gavage (300 mg/kg) with the pure anthocyanins, delphinidin-3-O-glucoside and malvidin-3-O-glucoside, formulated with Labrasol, showed that malvidin-3-O-glucoside was significantly hypoglycemic while delphinidin-3-O-glucoside was not.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72813
Oenin chloride, ≥90% (HPLC)
C23H25ClO12