EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Maternal exposure to anti-androgenic compounds, vinclozolin, flutamide and procymidone, has no effects on spermatogenesis and DNA methylation in male rats of subsequent generations.
PMID 19303894
Abstract
To verify whether anti-androgens cause transgenerational effects on spermatogenesis and DNA methylation in rats, gravid Crl:CD(SD) female rats (4 or 5/group, gestational day (GD) 0=day sperm detected) were intraperitoneally treated with anti-androgenic compounds, such as vinclozolin (100 mg/kg/day), procymidone (100 mg/kg/day), or flutamide (10 mg/kg/day), from GD 8 to GD 15. Testes were collected from F1 male pups at postnatal day (PND) 6 for DNA methylation analysis of the region (210 bp including 7 CpG sites) within the lysophospholipase gene by bisulfite DNA sequencing method. F0 and F1 males underwent the sperm analysis (count, motility and morphology), followed by DNA methylation analysis of the sperm. Remaining F1 males were cohabited with untreated-females to obtain F2 male pups for subsequent DNA methylation analysis of the testes at PND 6. These analyses showed no effects on spermatogenesis and fertility in F1 males of any treatment group. DNA methylation status in testes (F1 and F2 pups at PND 6) or sperms (F1 males at 13 weeks old) of the treatment groups were comparable to the control at all observation points, although DNA methylation rates in testes were slightly lower than those in sperm. In F0 males, no abnormalities in the spermatogenesis, fertility and DNA methylation status of sperm were observed. No transgenerational abnormalities of spermatogenesis and DNA methylation status caused by anti-androgenic compounds were observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36640
Procymidone, PESTANAL®, analytical standard
C13H11Cl2NO2