EMAIL THIS PAGE TO A FRIEND
Journal of Reproduction and Development
Mouse androgenetic embryonic stem cells differentiated to multiple cell lineages in three embryonic germ layers in vitro.
PMID 19305126