EMAIL THIS PAGE TO A FRIEND
Journal of protein chemistry
Effect of boronic acids on antifreeze proteins.
PMID 1930631
Abstract
The activity of antifreeze glycoprotein from the blood serum of Boreagadus saida was strongly inhibited by ions of organic boronic acids as well as by borate. The activity of nonglycoprotein from the blood serum of Pseudopleuronectus americanus, however, was not similarly inhibited. The inhibition by borate is thus specific for molecules with the carbohydrate moiety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
325104
3-Nitrophenylboronic acid, ≥97%
C6H6BNO4