EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
Enzyme-enhanced fluorescence detection of DNA on etched optical fibers.
PMID 19307112
Abstract
A novel DNA biosensor based on enzyme-enhanced fluorescence detection on etched optical fibers was developed. The hybridization complex of DNA probe and biotinylated target was formed on the etched optical fiber, and was then bound with streptavidin labeled horseradish peroxidase (streptavidin-HRP). The target DNA was quantified through the fluorescent detection of bi-p,p'-4-hydroxyphenylacetic acid (DBDA) generated from the substrate 4-hydroxyphenylacetic acid (p-HPA) under the catalysis of HRP, with a detection limit of 1 pM and a linear range from 1.69 pM to 169 pM. It is facile to regenerate this sensor through surface treatment with concentrated urea solution. It was discovered that the sensor can retain 70% of its original activity after three detection-regeneration cycles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H50004
4-Hydroxyphenylacetic acid, 98%
C8H8O3